Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SR-BI Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 98 publications
$268.00 - $584.50
Specifications
Antigen | SR-BI |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000-1:5000, Simple Western 1:100, Flow Cytometry 1:400. Use reported in scientific literature (PMID 23029167), Immunohistochemistry 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 24244566), Immunoblotting reported in scientific literature (PMID 27599291), Flow (Intracellular), Knockout Validated, Block/Neutralize reported in scientific literature (PMID 12119305; 26905520) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400101SS
![]() |
Novus Biologicals
NB400101SS |
0.025 mL |
Each for $268.00
|
|
|||||
NB400101
![]() |
Novus Biologicals
NB400101 |
0.1 mL |
Each for $584.50
|
|
|||||
Description
SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Bovine, Chinese Hamster, Mustelid, Primate, S. japonicum samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated, Block/Neutralize.Specifications
SR-BI | |
Western Blot 1:1000-1:5000, Simple Western 1:100, Flow Cytometry 1:400. Use reported in scientific literature (PMID 23029167), Immunohistochemistry 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 24244566), Immunoblotting reported in scientific literature (PMID 27599291), Flow (Intracellular), Knockout Validated, Block/Neutralize reported in scientific literature (PMID 12119305; 26905520) | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
SCARB1 | |
IgG | |
Affinity Purified | |
Human, mouse, mink, primate, rat, Chinese Hamster and Schistosoma japonicum. bovine reactivity reported in scientific literature (PMID: 24196350). |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine, Chinese Hamster, Mustelid, Primate, S. japonicum | |
Q8WTV0, Q61009, P97943 | |
949 | |
A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [Uniprot: Q61009] | |
Primary | |
Store at 4C. Do not freeze. | |
82 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title