Learn More
Description
Specifications
Specifications
| Antigen | SR-BI/SR-BII |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Neutralization |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:400. Use reported in scientific literature (PMID 22097902), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 5 ug/ml, Knockout Validated, Block/Neutralize reported in scientific literature (PMID 24859737) |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Accession No. | Q61009, O35114 |
| Gene Alias | CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1CD36 antigen-like 1, CLA-1HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, scavenger receptor class B type III, scavenger receptor class B, member 1, SRB1scavenger receptor class B member 1, SR-BIMGC138242 |
| Gene Symbols | SCARB1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
