Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR-BI/SR-BII Antibody, Novus Biologicals™
SDP

Catalog No. NB400134SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400134SS 0.025 mL
NB400134 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400134SS Supplier Novus Biologicals Supplier No. NB400134SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

SR-BI/SR-BII Polyclonal specifically detects SR-BI/SR-BII in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative), Knockout Validated, Block/Neutralize.

Specifications

Antigen SR-BI/SR-BII
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Neutralization
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:400. Use reported in scientific literature (PMID 22097902), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 5 ug/ml, Knockout Validated, Block/Neutralize reported in scientific literature (PMID 24859737)
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. Q61009, O35114
Gene Alias CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1CD36 antigen-like 1, CLA-1HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, scavenger receptor class B type III, scavenger receptor class B, member 1, SRB1scavenger receptor class B member 1, SR-BIMGC138242
Gene Symbols SCARB1
Host Species Rabbit
Immunogen A peptide from the extracellular domain (residues 230-380) of Scavenger Receptor-BI/BII that was expressed as two tandem copies in bacteria using the pET system.
Molecular Weight of Antigen 82 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 949
Test Specificity Mouse. Previous lots have been shown to react with rat and human as well.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.