Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SRBC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | SRBC |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2539
![]() |
Bethyl Laboratories, Inc
A302419AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein kinase C delta (PKC-d) binding protein (PRKCDBP), also known as cavin-3, is a member of the cavin family of proteins that are involved in caveolin formation and regulation. PRKCDBP was initially identified in a screen of cultured cell lines for proteins that were strongly induced by serum starvation. Studies indicate that PRKCDBP binds not only to PKC-delta but also to caveolin-1 and helps regulate caveolin traffic and function. Similar to other members of the cavin family, the expression of PRKCDBP was found to be down-regulated in various cancer cell lines, suggesting a possible tumor suppressor function of PRKCDBP.Specifications
SRBC | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
PRKCDBP; protein kinase C, delta binding protein; HSRBC; MGC20400; SRBC; sdr-related gene product that binds to c-kinase; cavin-3 | |
CAVIN3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q969G5 | |
112464 | |
Between 250 and 261 | |
Primary | |
4° C | |
CAVIN3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title