Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100312 Supplier Novus Biologicals Supplier No. NB100312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRC1 Polyclonal specifically detects SRC1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen SRC1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry, Immunoprecipitation
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1
Gene Symbols NCOA1
Host Species Rabbit
Immunogen A 29 amino acid synthetic peptide, which represented a portion of human nuclear receptor coactivator 1 encoded within exon 12 (LocusLink ID 8648).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8648
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.