Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300541 Supplier Novus Biologicals Supplier No. NB300541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

SRC1 Polyclonal specifically detects SRC1 in Human, Mouse, Rat, Amphibian, Avian samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen SRC1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation, Immunohistochemistry-Frozen 1 ug/ml
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. Q15788
Gene Alias bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1
Gene Symbols NCOA1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(8) S D P A N P D S H K R K G S P C(24) of human SRC-1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8648
Test Specificity Detects steroid receptor coactivator-1 (SRC-1).
Target Species Human, Mouse, Rat, Amphibian, Avian
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.