Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292191 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292191 Supplier Novus Biologicals Supplier No. NB100313PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRC1 Polyclonal specifically detects SRC1 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen SRC1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1400 and the C-terminus (residue 1441) of human Nuclear Receptor Coactivator 1 using the numbering given in entry NP_003734.3 (GeneID 8648).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8648
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.