Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRCAP Antibody, Novus Biologicals™
SDP

Catalog No. NBP276468 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276468 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276468 Supplier Novus Biologicals Supplier No. NBP276468
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SRCAP Polyclonal specifically detects SRCAP in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA, Immunofluorescence.

Specifications

Antigen SRCAP
Applications ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000, Immunofluorescence
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias domino homolog 1, Domino homolog 2, DOMO1, EAF1, EC 3.6.1, EC 3.6.4.-, FLJ44499, KIAA0309helicase SRCAP, Snf2-related CBP activator, Snf2-related CREBBP activator protein, Swi2/Snf2-related ATPase homolog, domino homolog 1, SWR1Snf2-related CBP activator protein
Gene Symbols SRCAP
Host Species Goat
Immunogen Peptide with sequence C-DEAQNIKNFKSQR corresponding to internal region according to NP_006653.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 10847.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.