Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRCAP Antibody, Novus Biologicals™
SDP

Catalog No. NB096382 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096382 Supplier Novus Biologicals Supplier No. NBP305265
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRCAP Polyclonal specifically detects SRCAP in Human samples. It is validated for Western Blot.

Specifications

Antigen SRCAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias domino homolog 1, Domino homolog 2, DOMO1, EAF1, EC 3.6.1, EC 3.6.4.-, FLJ44499, KIAA0309helicase SRCAP, Snf2-related CBP activator, Snf2-related CREBBP activator protein, Swi2/Snf2-related ATPase homolog, domino homolog 1, SWR1Snf2-related CBP activator protein
Host Species Rabbit
Immunogen Recombinant protein of human SRCAP.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10847
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.