Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRD5A2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300981 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300981 Supplier Novus Biologicals Supplier No. NB300981
Add to Cart
Add to Cart

Hamster Polyclonal Antibody

SRD5A2 Polyclonal specifically detects SRD5A2 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen SRD5A2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3-1 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000339.
Gene Alias 3-oxo-5-alpha-steroid 4-dehydrogenase 2,5 alpha-SR2, EC 1.3.99.5, MGC138457, S5AR 2, SR type 2, Steroid 5-alpha-reductase 2,3-oxo-5 alpha-steroid 4-dehydrogenase 2, steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta4-dehydrogenase alpha 2), Type II 5-alpha reductase
Gene Symbols SRD5A2
Host Species Hamster
Immunogen Peptide with sequence QVQCQQSPVLAG corresponding to N-Terminus according to NP_000339.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6716
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.