Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Antibody (2A4), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB60582A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60582A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60582A532 Supplier Novus Biologicals Supplier No. NB600582AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SREBP1 Monoclonal specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen SREBP1
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone 2A4
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
Gene Symbols SREBF1
Host Species Mouse
Immunogen 6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProtNo. P36956]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6720
Target Species Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.