Learn More
Description
Specifications
Specifications
| Antigen | SREBP1 |
| Applications | Western Blot, Immunohistochemistry, Immunoassay |
| Classification | Monoclonal |
| Clone | 2A4 |
| Conjugate | DyLight 488 |
| Dilution | Western Blot, Immunohistochemistry, Knockout Validated |
| Gene Accession No. | P36956 |
| Gene Alias | BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 |
| Gene Symbols | SREBF1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
