Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Antibody (2A4), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB354809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB354809 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB354809 Supplier Novus Biologicals Supplier No. NB600582MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SREBP1 Monoclonal antibody specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Western Blot, Immunohistochemistry (Frozen), KnockDown

Specifications

Antigen SREBP1
Applications Western Blot, Simple Western, Immunohistochemistry (Frozen), KnockDown
Classification Monoclonal
Clone 2A4
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Immunohistochemistry-Frozen, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
Host Species Mouse
Immunogen 6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProt# P36956]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6720
Target Species Human, Mouse, Rat, Canine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.