Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1002215SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002215SS 0.025 mL
NB1002215 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002215SS Supplier Novus Biologicals Supplier No. NB1002215SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 29 publications

SREBP1 Polyclonal specifically detects SREBP1 in Bovine, Hamster, Human, Mouse, Ovine, Pig, Plant, Rat, Staphylococcus epidermidis samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Simple Western, TMA, Western Blot.

Specifications

Antigen SREBP1
Applications Array, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Simple Western 1:25, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200 - 1:500, Microarray reported in scientific literature (PMID 33278777)
Formulation Tris-Glycine, 0.15 M NaCl with 0.02% Sodium Azide
Gene Accession No. P36956
Gene Alias BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
Gene Symbols SREBF1
Host Species Rabbit
Immunogen A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 300-400). [Uniprot: P36956]
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6720
Test Specificity Human, Mouse, Rat and Porcine. Immunogen has 94% homology to goat, bovine, and sheep.
Target Species Bovine, Mouse, Hamster, Human, Mouse, Plant, Ovine, Pig, Plant, Rat, Staphylococcus epidermidis
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.