Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 29 publications

$215.16 - $578.99

Specifications

Antigen SREBP1
Concentration 1.0 mg/mL
Dilution Western Blot 1:500, Simple Western 1:25, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200 - 1:500, Microarray reported in scientific literature (PMID 33278777)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002215SS
SDP
View Documents
Novus Biologicals
NB1002215SS
0.025 mL
Each for $215.16
Only null left
Add to Cart
 
NB1002215
SDP
View Documents
Novus Biologicals
NB1002215
0.1 mL
Each for $578.99
Only null left
Add to Cart
 
Description

Description

SREBP1 Polyclonal specifically detects SREBP1 in Human, Mouse, Rat, Porcine, Bovine, Hamster, Plant samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.
Specifications

Specifications

SREBP1
Western Blot 1:500, Simple Western 1:25, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200 - 1:500, Microarray reported in scientific literature (PMID 33278777)
Polyclonal
Rabbit
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Tris-Glycine, 0.15 M NaCl with 0.02% Sodium Azide
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
SREBF1
IgG
Affinity Purified
Human, Mouse, Rat and Porcine. Immunogen has 94% homology to goat, bovine, and sheep.
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
RUO
Human, Mouse, Rat, Pig, Bovine, Plant
P36956
6720
A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 300-400). [Uniprot: P36956]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
65 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.