Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB355107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355107 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355107 Supplier Novus Biologicals Supplier No. NB1002215PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SREBP1 Polyclonal specifically detects SREBP1 in Bovine, Hamster, Human, Mouse, Pig, Plant, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Western Blot.

Specifications

Antigen SREBP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Microarray
Formulation PBS
Gene Alias BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
Host Species Rabbit
Immunogen A synthetic peptide made to a portion of the human SREBP1 protein sequence (between residues 300-400). [Uniprot: P36956]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6720
Target Species Bovine, Hamster, Human, Mouse, Pig, Plant, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.