Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP2 Antibody (1D2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP154446SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154446SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154446SS Supplier Novus Biologicals Supplier No. NBP154446SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 15 publications

SREBP2 Monoclonal specifically detects SREBP2 in Human, Chicken samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays.

Specifications

Antigen SREBP2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Gel Shift
Classification Monoclonal
Clone 1D2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 5 ug/mL, ELISA, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunoprecipitation 10 ug/mg protein, Gel Super Shift Assays
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q12772
Gene Alias BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2
Gene Symbols SREBF2
Host Species Mouse
Immunogen Human SREBP2 protein (amino acids 48-403). [UniProt# Q12772]
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6721
Test Specificity Recognizes the precursor and mature forms of human SREBP2.
Target Species Human, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.