Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB264938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB264938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB264938 Supplier Novus Biologicals Supplier No. NBP171880B

Rabbit Polyclonal Antibody

SREBP2 Polyclonal antibody specifically detects SREBP2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot

Specifications

Antigen SREBP2
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human Sterol Regulatory Element Binding Transcription Factor 2 using the numbering given in entry NP_004590.2 (GeneID 6721).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6721
Target Species Human, Mouse, Zebrafish
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.