Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREC-I/SCARF1 Antibody (373606), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB365213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365213 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365213 Supplier Novus Biologicals Supplier No. FAB2409Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SREC-I/SCARF1 Monoclonal antibody specifically detects SREC-I/SCARF1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen SREC-I/SCARF1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 373606
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Acetyl LDL receptor, KIAA0149endothelial cells scavenger receptor, MGC47738, scavenger receptor class F, member 1, Scavenger receptor expressed by endothelial cells 1, SREC1, SREC-I, SRECscavenger receptor expressed by endothelial cells
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SREC-I/SCARF1, Ser20-Thr421, Accession # Q14162
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8578
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.