Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRI Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB328945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328945 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328945 Supplier Novus Biologicals Supplier No. NBP300310MFV610

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

SRI Polyclonal antibody specifically detects SRI in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SRI
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias calcium binding protein amplified in mutlidrug-resistant cells, CP22, CP-22, FLJ26259,22 kDa protein, H_RG167B05.1, SCN, sorcin, V19
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human SRI (Accession#: P30626; Met1-Val198)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 6717
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.