Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRP14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132034 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132034 Supplier Novus Biologicals Supplier No. NBP132034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRP14 Polyclonal specifically detects SRP14 in Human samples. It is validated for Western Blot.

Specifications

Antigen SRP14
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Alias ALURBP, MGC14326,18 kDa Alu RNA-binding protein, signal recognition particle 14 kDa protein, signal recognition particle 14kD (homologous Alu RNA-binding protein), signal recognition particle 14kDa (homologous Alu RNA binding protein)
Gene Symbols SRP14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SRP14. The exact sequence is proprietary.
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6727
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identitiy: Rhesus Monkey (100%). Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.