Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRPK3/STK23 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220515 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220515 Supplier Novus Biologicals Supplier No. NBP220515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRPK3/STK23 Polyclonal specifically detects SRPK3/STK23 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SRPK3/STK23
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UPE1
Gene Alias MGC102944, MSSK-1, MSSK1EC 2.7.11.1, Muscle-specific serine kinase 1, serine arginine rich protein-specific kinase 3, Serine/arginine-rich protein-specific kinase 3, serine/threonine kinase 23, Serine/threonine-protein kinase 23, serine/threonine-protein kinase SRPK3, SFRS protein kinase 3, SR-protein-specific kinase 3, SRSF protein kinase 3, STK23
Gene Symbols SRPK3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STK23. The exact sequence is proprietary.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 26576
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.