Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRPRB Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SRPRB
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159710
SDP
View Documents
Novus Biologicals
NBP159710
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

SRPRB Polyclonal specifically detects SRPRB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SRPRB
Unconjugated
RUO
APMCF1, Protein APMCF1, signal recognition particle receptor subunit beta, signal recognition particle receptor, B subunit, signal recognition particle receptor, beta subunit, SR-beta
SRPRB
IgG
This product is specific to Subunit or Isoform: beta.
Polyclonal
Rabbit
Q9Y5M8
58477
Synthetic peptides corresponding to SRPRB(signal recognition particle receptor, B subunit) The peptide sequence was selected from the middle region of SRPRB. Peptide sequence QDIAMAKSAKLIQQQLEKELNTLRVTRSAAPSTLYSSSTAPAQLGKKGKE.
Primary
30 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.