Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRPRB Antibody (OTI2D4) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP274351 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274351 Supplier Novus Biologicals Supplier No. NBP274351

Mouse Monoclonal Antibody

SRPRB Monoclonal specifically detects SRPRB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SRPRB
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D4
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:100
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias APMCF1, Protein APMCF1, signal recognition particle receptor subunit beta, signal recognition particle receptor, B subunit, signal recognition particle receptor, beta subunit, SR-beta
Gene Symbols SRPRB
Host Species Mouse
Immunogen Full length human recombinant protein of human SRPRB(NP_067026) produced in HEK293T cell.
Molecular Weight of Antigen 29.5 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 58477
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.