Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRPX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106437
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5106437 100 μL
1 options

Catalog No. PIPA5106437

Supplier: Invitrogen™ PA5106437

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SRPX2.

Sushi-repeat-containing protein X-linked 2 (SRPX2) is a neural gene functioning in the speech and language center of the human brain; mutations in this gene lead to epilepsy, speech dyspraxia, mental retardation and cognitive disorders. Recently, SRPX2 was found to be a novel mediator of angiogenesis and can act as a ligand for the urokinase-type plasminogen activator, a protein that can facilitate invasive migration of sprouting endothelial cells. SRPX2 is also overexpressed in gastric cancer, leading to increased phosphorylation levels of focal adhesion kinase and enhanced cellular migration and adhesion, suggesting that SRPX2 may be a potential target in the treatment of metastatic cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRPX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRPX2
Gene Accession No. B5DF94, O60687, Q8R054
Gene Alias 1110039C07Rik; BPP; CBPS; EMBL:AAI68975.1, ECO:0000312; PMGX; PubMed:24179158}; RESDX; RGD:1562444}; RGD1562444; RP11-524D16__A.1; SRP; SRPUL; SRPX2; srpx2 {ECO:0000312; sushi repeat containing protein X-linked 2; sushi repeat containing protein, X-linked 2; sushi repeat-containing protein SRPX2; sushi repeat-containing protein SRPX2 {ECO:0000303; sushi-repeat containing protein; sushi-repeat containing protein, X-linked 2; sushi-repeat protein; sushi-repeat protein upregulated in leukemia; sushi-repeat protein up-regulated in leukemia; sushi-repeat-containing protein, X-linked 2
Gene Symbols SRPX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRPX2(Accession O60687), corresponding to amino acid residues G131-P181.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27286, 317181, 68792
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.