Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SS18 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB320606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320606 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320606 Supplier Novus Biologicals Supplier No. NBP298476PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SS18 Polyclonal antibody specifically detects SS18 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SS18
Applications Immunocytochemistry
Classification Polyclonal
Conjugate PE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias MGC116875, Protein SYT, SSXTsynovial sarcoma, translocated to X chromosome, Synovial sarcoma translocated to X chromosome protein, synovial sarcoma translocation, chromosome 18, SYTprotein SSXT
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SS18 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6760
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.