Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSBIP1/SOSS-C Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB150229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB150229 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB150229 Supplier Novus Biologicals Supplier No. NBP298143AF750

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

SSBIP1/SOSS-C Polyclonal specifically detects SSBIP1/SOSS-C in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SSBIP1/SOSS-C
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias chromosome 9 open reading frame 80, HSPC043, hSSB-interacting protein 1, hSSBIP1, minute INTS3/hSSB-associated element, MISE, Sensor of single-strand DNA complex subunit C, Sensor of ssDNA subunit C, Single-stranded DNA-binding protein-interacting protein 1, SOSS complex subunit C, SOSSC, SOSS-C, SSB-interacting protein 1, SSBIP1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SSBIP1/SOSS-C fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58493
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.