Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSBP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB126729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB126729 100 μg
NB126730 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB126729 Supplier Novus Biologicals Supplier No. NBP310915100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSBP2 Polyclonal specifically detects SSBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SSBP2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias DKFZp686F03273, HSPC116, Sequence-specific single-stranded-DNA-binding protein 2, single-stranded DNA binding protein 2, single-stranded DNA-binding protein 2, SOSS-B2, SSDP2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human SSBP2 (NP_036578). Peptide sequence YPGGPRPPLRIPNQALGGVPGSQPLLPSGMDPTRQQGHPNMGGPMQRMTP
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23635
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.