Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-1 Antibody (FUT4/1478R), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB029922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029922 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB029922 Supplier Novus Biologicals Supplier No. NBP249879APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen SSEA-1
Applications Flow Cytometry
Classification Monoclonal
Clone FUT4/1478R
Conjugate APC
Formulation PBS
Gene Alias alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Host Species Rabbit
Immunogen Recombinant human full-length SSEA-1 protein (Uniprot: P22083)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2526
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.