Learn More
Description
Specifications
Specifications
| Antigen | SSEA-1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | FUT4/1478R |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant human full-length SSEA-1 protein (Uniprot: P22083) |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
