Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-1 Antibody (MEM-158), Novus Biologicals™
SDP

Catalog No. NB500376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500376 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500376 Supplier Novus Biologicals Supplier No. NB500376
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSEA-1 Monoclonal specifically detects SSEA-1 in Human, Bovine (Negative), Porcine (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen CD15/Lewis X
Applications Flow Cytometry, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone MEM-158
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 to 4 μg/mL, Immunoprecipitation 1:50, CyTOF-ready
Gene Alias CD 15, CD-15, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Gene Symbols FUT4
Host Species Mouse
Immunogen Human granulocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Embryonic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2526
Test Specificity The antibody MEM-158 reacts with CD15, a cell membrane molecule 3-fucosyl-N-acetyllactosamine (3-FAL) strongly expressed on granulocytes, monocytes, macrophages, mast cells; it is also present on Langerhans cells and some myeloid precursors cells. HLDA VI; WS Code AS A053
Target Species Human, Bovine (Negative), Porcine (Negative), Sheep (Negative)
Content And Storage Store at 4C. Do not freeze.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.