Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-3 Antibody (MC-631), Novus Biologicals™
SDP

Catalog No. NB1001832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001832 0.1 mL
NB1001832SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001832 Supplier Novus Biologicals Supplier No. NB1001832
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 7 publications

SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SSEA-3
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MC-631
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 10-20 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Gene Alias Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3
Host Species Rat
Immunogen 4-8 cell stage mouse embryos.
Purification Method IgM purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Test Specificity Recognizes a carbohydrate epitope of SSEA-3
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.