Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-3 Antibody (MC-631), Novus Biologicals™
SDP

Rat Monoclonal Antibody has been used in 7 publications

$206.50 - $499.50

Specifications

Antigen SSEA-3
Clone MC-631
Concentration 1 mg/mL
Dilution Western Blot 1:500, Flow Cytometry 10-20 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001832SS
SDP
View Documents
Novus Biologicals
NB1001832SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB1001832
SDP
View Documents
Novus Biologicals
NB1001832
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications

Specifications

SSEA-3
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rat
Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers
Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3
IgM
IgM purified
Recognizes a carbohydrate epitope of SSEA-3
MC-631
Western Blot 1:500, Flow Cytometry 10-20 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse
4-8 cell stage mouse embryos.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.