Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-4 Antibody (MC-813-70), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP226644Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226644Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226644Y Supplier Novus Biologicals Supplier No. NBP226644AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse, Canine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).

Specifications

Antigen SSEA-4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone MC-813-70
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
Gene Accession No. NP_803131
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Chicken, Dog, Human, Mouse, New World Monkey, Rabbit.
Target Species Human, Mouse, Canine, Chicken, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.