Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-4 Antibody (MC-813-70), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB304916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304916 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304916 Supplier Novus Biologicals Supplier No. FAB1435MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen SSEA-4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone MC-813-70
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias SSEA-4,stage-specific embryonic antigen 4
Host Species Mouse
Immunogen 2120Ep human embryonal carcinoma cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.