Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA-4 Mouse anti-Canine, Chicken, Human, Mouse, Primate, Rabbit, DyLight 755, Clone: MC-813-70, Novus Biologicals™
SDP

Catalog No. NB226644IR1 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB226644IR1 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB226644IR1 Supplier Novus Biologicals Supplier No. NBP226644IR0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSEA-4 Monoclonal specifically detects SSEA-4 in Human, Mouse, Canine, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).

Specifications

Antigen SSEA-4
Applications Flow Cytometry
Classification Monoclonal
Clone MC-813-70
Conjugate DyLight 755
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. NP_803131
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Canine, Chicken, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.