Learn More
SSEA4 Monoclonal Antibody (eBioMC-813-70 (MC-813-70)), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$147.00 - $371.25
Specifications
Antigen | SSEA4 |
---|---|
Clone | eBioMC-813-70 (MC-813-70) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-884-380
![]() |
Invitrogen™
50884380 |
25 μg |
$196.00
$147.00 / Each Save $49.00 25% Off |
|
|||||
50-884-382
![]() ![]() |
Invitrogen™
50884382 |
100 μg |
$495.00
$371.25 / Each Save $123.75 25% Off |
![]() |
|||||
Description
The eBioMC-813-70 MAb reacts with a glycolipid carbohydrate epitope on Stage-specific embryonic antigen-4 (SSEA-4) protein. Expression is found on human teratocarcinoma stem cells (EC), human embryonic germ cells (EG), human induced pluripotent stem (iPS) cells, and human embryonic stem cells (ES) with a decrease in expression upon differentiation. Recently expression has also been found on bone marrow mesenchymal stem cells. The eBioMC-813-70 antibody has been reported to crossreact to mouse, rabbit, canine, chicken, and Old and New World monkeys.
SSEA-4 (Stage-specific embryonic antigen 4) is a cell surface antigen, expressed along with SSEA-3, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA-1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.Specifications
SSEA4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG3 | |
Affinity chromatography | |
Antibody |
eBioMC-813-70 (MC-813-70) | |
Flow Cytometry, Immunocytochemistry | |
eFluor 660 | |
Mouse | |
Human, Mouse, Canine, Rabbit, Monkey, Chicken | |
SSEA-4; stage-specific embryonic antigen 4 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.