Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSEA5 Monoclonal Antibody (8E11), DyLight™ 755, Invitrogen™

Catalog No. MA1144D755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1144D755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1144D755 Supplier Thermo Scientific Supplier No. MA1144D755

Mouse Monoclonal Antibody

Immunofluorescent analysis of MA1-144-D755 shows positive staining of SSEA-5 in human embryonal carcinoma NCCIT cells.

SSEA5 Monoclonal antibody specifically detects SSEA5 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA5
Applications Immunocytochemistry
Classification Monoclonal
Clone 8E11
Concentration 1 mg/mL
Conjugate DyLight 755
Formulation PBS with proprietary stabilizer and 0.02% sodium azide
Gene Alias SSEA-5
Host Species Mouse
Immunogen Undifferentiated H9 human embryonic stem cells
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.