Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSH1, p Ser978 Antibody, Novus Biologicals™
SDP

Catalog No. NB387842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387842 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387842 Supplier Novus Biologicals Supplier No. NBP323411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSH1 Polyclonal antibody specifically detects SSH1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SSH1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:100
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias EC 3.1.3.16, EC 3.1.3.48, FLJ21928, hSSH-1L, KIAA1298FLJ38102, protein phosphatase Slingshot homolog 1, slingshot 1, slingshot homolog 1 (Drosophila), SSH1L, SSH-1L, SSH-like protein 1
Host Species Rabbit
Immunogen SSH1 (Ser-978) antibody was generated from a phospho-peptide that included amino acids surrounding serine 978 in human SSH1. This sequence has high homology to the conserved site in rat, mouse, chicken, and zebrafish SSH1L. The sequence also has high homology to the conserved site in SSH2L (Ser-1259). [UniProt# Q8WYL5]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 54434
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.