Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSH1 Rabbit anti-Human, Mouse, Rat, Monkey, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB018655 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018655 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018655 Supplier Novus Biologicals Supplier No. NBP1506360.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSH1 Polyclonal specifically detects SSH1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SSH1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.69 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000, ELISA 1:10000
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%.
Gene Accession No. NP_061857
Gene Alias EC 3.1.3.16, EC 3.1.3.48, FLJ21928, hSSH-1L, KIAA1298FLJ38102, protein phosphatase Slingshot homolog 1, slingshot 1, slingshot homolog 1 (Drosophila), SSH1L, SSH-1L, SSH-like protein 1
Host Species Rabbit
Immunogen Phosphorylated synthetic peptide corresponding to unique amino acid sequence on Slingshot homolog 1 protein.
Purification Method Immunogen affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 54434
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.