Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSH3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060674 Supplier Novus Biologicals Supplier No. NB10060674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSH3 Polyclonal specifically detects SSH3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen SSH3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 5 ug/mg lysate, Immunohistochemistry-Paraffin, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060327.2
Gene Alias EC 3.1.3.16, EC 3.1.3.48, FLJ10928, FLJ20515, FLJ42240, hSSH-3L, protein phosphatase Slingshot homolog 3, slingshot 3, slingshot homolog 3 (Drosophila), SSH3, SSH3L, SSH-3L, SSH-like protein 3
Gene Symbols SSH3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human slingshot homolog 3 using the numbering given in entry NP_060327.2 (GeneID 54961).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54961
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.