Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ssPA/Stringent Starvation Protein A Antibody (113), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB070635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070635 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070635 Supplier Novus Biologicals Supplier No. NBP290173AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ssPA/Stringent Starvation Protein A Monoclonal specifically detects ssPA/Stringent Starvation Protein A in E. coli samples. It is validated for ELISA.

Specifications

Antigen ssPA/Stringent Starvation Protein A
Applications ELISA
Classification Monoclonal
Clone 113
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ECK3218, Pog, ssp, stringent starvation protein A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant E. coli ssPA/Stringent Starvation Protein A (P0ACA3; Met1-Ser212).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944744
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.