Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SSR 180711 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
SSR 180711 hydrochloride is a partial agonist at α7 nAChR. Enhances episodic memory and reverses MK-801 -induced deficits in retention of episodic memory. Increases extracellular dopamine levels in rat prefrontal cortex. Also exhibits antidepressant-like properties.
Specifications
Specifications
Chemical Name or Material | 4-Bromophenyl 1,4-diazabicyclo[3.2.2]nonane-4-carboxylate hydrochloride |
CAS | 446031-79-4 |
Quantity | 10 mg |
Target | Nicotinic (a7) Receptor Agonists |
Molecular Formula | C14H17BrN2O2.HCl |
Formula Weight | 361.66 |
Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction