Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSR2 Antibody (31G6), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309957 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309957 Supplier Novus Biologicals Supplier No. NBP242647MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSR2 Monoclonal antibody specifically detects SSR2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SSR2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 31G6
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp686F19123, Signal sequence receptor subunit beta, signal sequence receptor, beta (translocon-associated protein beta), SSR-beta, TLAP, translocon-associated protein beta, translocon-associated protein subunit beta, TRAP-beta, TRAPBTRAP-BETA
Host Species Mouse
Immunogen Recombinant human SSR2 (18-149aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6746
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.