Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSR3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB097487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097487 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097487 Supplier Novus Biologicals Supplier No. NBP297569R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSR3 Polyclonal specifically detects SSR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SSR3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias signal sequence receptor, gamma (translocon-associated protein gamma), SSR gamma, SSR-gamma, translocon-associated protein gamma subunit, translocon-associated protein subunit gamma, TRAP-complex gamma subunit, TRAP-gamma, TRAPGSignal sequence receptor subunit gamma
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SSR3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6747
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.