Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSRP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170510 25 μg
NB170511 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170510 Supplier Novus Biologicals Supplier No. NBP31867625UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SSRP1 Polyclonal antibody specifically detects SSRP1 in Human samples. It is validated for Western Blot

Specifications

Antigen SSRP1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS
Gene Accession No. Q08945
Gene Alias Chromatin-specific transcription elongation factor 80 kDa subunit, cisplatin-DNA SSRP, facilitates chromatin remodeling 80 kDa subunit, Facilitates chromatin transcription complex 80 kDa subunit, Facilitates chromatin transcription complex subunit SSRP1, FACT, FACT complex subunit SSRP1, FACT80FACT 80 kDa subunit, FACTp80, high mobility group box, hSSRP1, Recombination signal sequence recognition protein 1, structure specific recognition protein 1, Structure-specific recognition protein 1, T160
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human SSRP1 (Structure-specific recognition protein 1), the GST tagged N-terminal part of the protein.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6749
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.