Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSSCA1 Antibody (OTI2F5), FITC, Novus Biologicals™
SDP

Catalog No. NBP274359F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274359F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274359F Supplier Novus Biologicals Supplier No. NBP274359F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSSCA1 Monoclonal specifically detects SSSCA1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SSSCA1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2F5
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias Autoantigen p27, centromeric autoantigen (27kD), p27, Sjoegren syndrome/scleroderma autoantigen 1, Sjogren syndrome/scleroderma autoantigen 1, Sjogren′s syndrome/scleroderma autoantigen 1
Gene Symbols ZNRD2
Host Species Mouse
Immunogen Full length recombinant protein of human SSSCA1 (NP_006387) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10534.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.