Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSTK-IP Antibody, Novus Biologicals™
SDP

Catalog No. NB397309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397309 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397309 Supplier Novus Biologicals Supplier No. NBP23392325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SSTK-IP Polyclonal antibody specifically detects SSTK-IP in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SSTK-IP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Accession No. Q96A04
Gene Alias C1orf182, chromosome 1 open reading frame 182, MGC26877, RP11-443G18.5, SIP, SSTK-interacting protein (SSTK-IP), SSTK-IP
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MERHTSHPNRKVPAKEEANAVPLCRAKPSPSYINLQASSPPATFLNIQTTKLPSVDHKPKECLG
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128229
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.