Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SSTK-IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | SSTK-IP |
---|---|
Dilution | Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SSTK-IP Polyclonal antibody specifically detects SSTK-IP in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
SSTK-IP | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q96A04 | |
128229 | |
IgG | |
Immunogen affinity purified |
Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2) and 40% Glycerol | |
C1orf182, chromosome 1 open reading frame 182, MGC26877, RP11-443G18.5, SIP, SSTK-interacting protein (SSTK-IP), SSTK-IP | |
This antibody was developed against a recombinant protein corresponding to amino acids: MERHTSHPNRKVPAKEEANAVPLCRAKPSPSYINLQASSPPATFLNIQTTKLPSVDHKPKECLG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title