Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ssu72 Antibody (45E2), PerCP, Novus Biologicals™
SDP

Catalog No. NB308708 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308708 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308708 Supplier Novus Biologicals Supplier No. NBP260233PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ssu72 Monoclonal antibody specifically detects Ssu72 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Ssu72
Applications Western Blot, ELISA
Classification Monoclonal
Clone 45E2
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias CTD phosphatase SSU72, EC 3.1.3.16, FLJ13048, HSPC182, PNAS-120, RNA polymerase II subunit A C-terminal domain phosphatase SSU72, SSU72 RNA polymerase II CTD phosphatase homolog (S. cerevisiae), Ssu72 RNA polymerase II CTD phosphatase homolog (yeast)
Host Species Mouse
Immunogen Recombinant human SSU72 (1-194aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 29101
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.